ContactUsLinkComponent
Hydrometer DIN 12791/BS 718, series S 50 SP precision, 0,800-0,850:0,001g/cm , without therm., ref. temp. 15°C, max. 190mm long, sui table for government verification 1 * 1 items
Catalog # AMARH811208
Supplier:  Amarell
Hydrometer DIN 12791/BS 718, series S 50 SP precision, 0,800-0,850:0,001g/cm , without therm., ref. temp. 15°C, max. 190mm long, sui table for government verification 1 * 1 items
Catalog # AMARH811208
Supplier:  Amarell
Frequently Bought Together