ContactUsLinkComponent
Thermometer similar to IP 62 C, solid stem, white backed, -5+300:1°C, gallium filled, durable pigment, suitable for government verif ication 1 * 1 items
  AMARA340830-GA
 :  Amarell
Thermometer similar to IP 62 C, solid stem, white backed, -5+300:1°C, gallium filled, durable pigment, suitable for government verif ication 1 * 1 items
  AMARA340830-GA
 :  Amarell
Frequently Bought Together