ContactUsLinkComponent
Fadentherm., 0+250:1°C, solid stem, white backed, blue special liquid, 360x6-7mm, bulb length 100mm, partial immersion, 16 B glass, with button at top, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARL44108-FL
Supplier:  Amarell
Fadentherm., 0+250:1°C, solid stem, white backed, blue special liquid, 360x6-7mm, bulb length 100mm, partial immersion, 16 B glass, with button at top, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARL44108-FL
Supplier:  Amarell
Frequently Bought Together