ContactUsLinkComponent
Flash point therm., similar Marcusson, similar DIN 12785, +190+410:1°C, 48mm immersion, 300x9-10mm, milk glass scale, cap. prismati c, colourless, gallium filled, Richter-top finish, spherical bulb, null
Catalog # AMARM234002-GA/VC
Supplier:  Amarell
Flash point therm., similar Marcusson, similar DIN 12785, +190+410:1°C, 48mm immersion, 300x9-10mm, milk glass scale, cap. prismati c, colourless, gallium filled, Richter-top finish, spherical bulb, null
Catalog # AMARM234002-GA/VC
Supplier:  Amarell
Frequently Bought Together