ContactUsLinkComponent
Flash point-Therm., similar Pensky-Martens, similar DIN 12785, +20+200:1C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prismatic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, null
Catalog # AMARM230042-FL/VC
Supplier:  Amarell
Flash point-Therm., similar Pensky-Martens, similar DIN 12785, +20+200:1C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prismatic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, null
Catalog # AMARM230042-FL/VC
Supplier:  Amarell
Frequently Bought Together