ContactUsLinkComponent
Thermometer similar to IP 47 C, solid stem, white backed, 155+170:0,5°C, gallium filled, total immersion, 155x5,5-7mm, durable pigme nt, suitable for government verification 1 * 1 items
Catalog # AMARA340670-CB
Supplier:  Amarell
Thermometer similar to IP 47 C, solid stem, white backed, 155+170:0,5°C, gallium filled, total immersion, 155x5,5-7mm, durable pigme nt, suitable for government verification 1 * 1 items
Catalog # AMARA340670-CB
Supplier:  Amarell
Frequently Bought Together