ContactUsLinkComponent
Hydrometer DIN 12791/BS 718, series L 50 SP precision, 0,900-0,950:0,0005g/ml, without therm., ref. temp. 15C, max. 335mm long, sui table for government verification 1 * 1 items
Catalog # AMARH806312
Supplier:  Amarell
Hydrometer DIN 12791/BS 718, series L 50 SP precision, 0,900-0,950:0,0005g/ml, without therm., ref. temp. 15C, max. 335mm long, sui table for government verification 1 * 1 items
Catalog # AMARH806312
Supplier:  Amarell
Frequently Bought Together