ContactUsLinkComponent
Cloud and pour point therm., DIN 12785, -80+20:1C, analogous to ASTM 6 C, solid stem, white backed, toluene, immersion 76mm, govern ment verified, with Verification Certificate with 4 test points 1 * 1 items
Catalog # AMARM242362-TOL/VC
Supplier:  Amarell
Cloud and pour point therm., DIN 12785, -80+20:1C, analogous to ASTM 6 C, solid stem, white backed, toluene, immersion 76mm, govern ment verified, with Verification Certificate with 4 test points 1 * 1 items
Catalog # AMARM242362-TOL/VC
Supplier:  Amarell
Frequently Bought Together