ContactUsLinkComponent
Cloud and pour point therm., similar DIN 12785, -38+50:1°C, similar ASTM 5 C, solid stem, white backed, red special liquid, immersio n 108mm, suitable for government verification 1 * 1 items
Catalog # AMARM242360-FL
Supplier:  Amarell
Cloud and pour point therm., similar DIN 12785, -38+50:1°C, similar ASTM 5 C, solid stem, white backed, red special liquid, immersio n 108mm, suitable for government verification 1 * 1 items
Catalog # AMARM242360-FL
Supplier:  Amarell
Frequently Bought Together