ContactUsLinkComponent
Viscometer-Therm., ähnlich Vogel-Ossag, ähnlich DIN 12785, +35+70:0,1°C, enclosed scale, capillary tube specially coated inside, wi th non-wetting blue special liquid, 470mm long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, null
Catalog # AMARM240104-CB
Supplier:  Amarell
Viscometer-Therm., ähnlich Vogel-Ossag, ähnlich DIN 12785, +35+70:0,1°C, enclosed scale, capillary tube specially coated inside, wi th non-wetting blue special liquid, 470mm long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, null
Catalog # AMARM240104-CB
Supplier:  Amarell
Frequently Bought Together