ContactUsLinkComponent
ASTM-Thermom., S116C, white backed, 18,9+25,1:0,01C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.625x7-8mm, durable pigment, null
Catalog # AMARA301550-CB
Supplier:  Amarell
ASTM-Thermom., S116C, white backed, 18,9+25,1:0,01C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.625x7-8mm, durable pigment, null
Catalog # AMARA301550-CB
Supplier:  Amarell
Frequently Bought Together