ContactUsLinkComponent
Fadentherm., 0+250:1°C, enclosed scale, cap. colourless, blue special liquid, 360xc.9mm, bulb length 100mm, partial immersion, 16 B glass, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARL44006-FL
Supplier:  Amarell
Fadentherm., 0+250:1°C, enclosed scale, cap. colourless, blue special liquid, 360xc.9mm, bulb length 100mm, partial immersion, 16 B glass, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARL44006-FL
Supplier:  Amarell
Frequently Bought Together