Orders LinkContactUsLinkComponent
Density hydrometers
Density hydrometers
Completely set with 25 instruments (00 to 22, incl. range finder), 300 mm
Catalog # AMARH800190
Supplier:   Amarell
Density hydrometers
Catalog # AMARH800190
Supplier:  Amarell
Density hydrometers
Density hydrometers
Completely set with 25 instruments (00 to 22, incl. range finder), 300 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
General purpose hydrometers for density areas.
  • Paper scale and shot weighted body
  • Hydrometer accuracy ±1 scale division
  • Thermometer inside the hydrometer body
Used in laboratories and industry.
Specifications
  • Description:
    Completely set with 25 instruments (00 to 22, incl. range finder)
  • Length:
    300 mm
  • Seq:
    88