ContactUsLinkComponent
Pocket therm., simple type, solid stem, -10+110:1C, 165x10mm, white backed, red special liquid filled, in yellow plastic case 1 * 1 items
Catalog # AMARG15656-FL
Supplier:  Amarell
Pocket therm., simple type, solid stem, -10+110:1C, 165x10mm, white backed, red special liquid filled, in yellow plastic case 1 * 1 items
Catalog # AMARG15656-FL
Supplier:  Amarell
Frequently Bought Together