ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 99 F, white backed, -58+41:0,5F, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 35mm, max.305x6,5-8mm, null
Catalog # AMARA301370-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 99 F, white backed, -58+41:0,5F, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 35mm, max.305x6,5-8mm, null
Catalog # AMARA301370-CB
Supplier:  Amarell
Frequently Bought Together