ContactUsLinkComponent
Precision therm., low temp., similar DIN, -38+20:0,1C, enclosed scale, cap. lens front, colourless, total immersion, capillary tub e specially coated inside, with non-wetting blue special liquid, 400x8-9mm, Richter-top finish, null
Catalog # AMARL33068-CB
Supplier:  Amarell
Precision therm., low temp., similar DIN, -38+20:0,1C, enclosed scale, cap. lens front, colourless, total immersion, capillary tub e specially coated inside, with non-wetting blue special liquid, 400x8-9mm, Richter-top finish, null
Catalog # AMARL33068-CB
Supplier:  Amarell
Frequently Bought Together