ContactUsLinkComponent
Distillation therm., similar to DIN 12779, 0+360:1°C, enclosed scale, cap. lens front, colourless, gallium filled, total immersion, length 375mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259612-GA
Supplier:  Amarell
Distillation therm., similar to DIN 12779, 0+360:1°C, enclosed scale, cap. lens front, colourless, gallium filled, total immersion, length 375mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259612-GA
Supplier:  Amarell
Frequently Bought Together