ContactUsLinkComponent
Thermometer similar to AFNOR, STL/1/380/620, +380+620:1°C, solid stem, frosted back, gallium filled, 375mm long, total immersion, wi th ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA411092-GA
Supplier:  Amarell
Thermometer similar to AFNOR, STL/1/380/620, +380+620:1°C, solid stem, frosted back, gallium filled, 375mm long, total immersion, wi th ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA411092-GA
Supplier:  Amarell
Frequently Bought Together