ContactUsLinkComponent
Thermometer similar to AFNOR, STL/1/180/420, +180+420:1°C, solid stem, white backed, gallium filled, 375mm long, total immersion, du rable pigment, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA411090-GA
Supplier:  Amarell
Thermometer similar to AFNOR, STL/1/180/420, +180+420:1°C, solid stem, white backed, gallium filled, 375mm long, total immersion, du rable pigment, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA411090-GA
Supplier:  Amarell
Frequently Bought Together