ContactUsLinkComponent
ASTM-Thermom., S56F, white backed, 66+95:0,05F, capillary tube specially coated inside, with non-wetting blue special liquid, tota l immersion, max.622x7-8mm, durable pigment, null
Catalog # AMARA300760-CB
Supplier:  Amarell
ASTM-Thermom., S56F, white backed, 66+95:0,05F, capillary tube specially coated inside, with non-wetting blue special liquid, tota l immersion, max.622x7-8mm, durable pigment, null
Catalog # AMARA300760-CB
Supplier:  Amarell
Frequently Bought Together