ContactUsLinkComponent
Hartong malt saccharimeter, 4-5:0,01%mas., with therm. +5+27:0,2°C with red or blue special liquid, with correction scale, reading b elow, ref. temp. 20°C, 400mm long 1 * 1 items
Catalog # AMARH847406-FL
Supplier:  Amarell
Hartong malt saccharimeter, 4-5:0,01%mas., with therm. +5+27:0,2°C with red or blue special liquid, with correction scale, reading b elow, ref. temp. 20°C, 400mm long 1 * 1 items
Catalog # AMARH847406-FL
Supplier:  Amarell
Frequently Bought Together