Thermometer similar to BS 593, A40C/total, 0+40:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, total immersion, durable pigment, null
Catalog # AMARA403202-CB
Supplier: Amarell
Frequently Bought Together