ContactUsLinkComponent
Twaddell Hydrometer, 102-138:0,5Twaddell, long type, without therm., ref. temp. 60F, 240mm long 1 * 1 items
Catalog # AMARH847128
Supplier:  Amarell
Twaddell Hydrometer, 102-138:0,5Twaddell, long type, without therm., ref. temp. 60F, 240mm long 1 * 1 items
Catalog # AMARH847128
Supplier:  Amarell
Frequently Bought Together