ContactUsLinkComponent
Thermometer similar to BS 1365, SA305C/80, 245+305:0,5°C, solid stem, white backed, gallium filled, 215mm long, immersion 80mm, dura ble pigment, suitable for government verification 1 * 1 items
Catalog # AMARA402530-GA
Supplier:  Amarell
Thermometer similar to BS 1365, SA305C/80, 245+305:0,5°C, solid stem, white backed, gallium filled, 215mm long, immersion 80mm, dura ble pigment, suitable for government verification 1 * 1 items
Catalog # AMARA402530-GA
Supplier:  Amarell
Frequently Bought Together