ContactUsLinkComponent
Hydrometer Baume, 50-70:1/1B, without therm., ref. temp. 15C, 240mm long 1 * 1 items
Catalog # AMARH842820
Supplier:  Amarell
Hydrometer Baume, 50-70:1/1B, without therm., ref. temp. 15C, 240mm long 1 * 1 items
Catalog # AMARH842820
Supplier:  Amarell
Frequently Bought Together