ContactUsLinkComponent
Thermometer similar to BS 1704, B/total, -35+30:0,5°C, white backed, capillary tube specially coated inside, with non-wetting blue special liquid total immersion, 300mm long, durable pigment, null
  AMARA402082-CB
 :  Amarell
Thermometer similar to BS 1704, B/total, -35+30:0,5°C, white backed, capillary tube specially coated inside, with non-wetting blue special liquid total immersion, 300mm long, durable pigment, null
  AMARA402082-CB
 :  Amarell
Frequently Bought Together