ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 22d, solid stem, white backed, 205+235:0,1°C, length 430mm, imm. 100mm, gallium filled, durable pigment, suitable for government calibration 1 * 1 items
Artikelnr. AMARA400170-GA
Lieferant:  Amarell
Thermometer similar to S.T.P.T.C. T 22d, solid stem, white backed, 205+235:0,1°C, length 430mm, imm. 100mm, gallium filled, durable pigment, suitable for government calibration 1 * 1 items
Artikelnr. AMARA400170-GA
Lieferant:  Amarell
Frequently Bought Together