Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Specific Gravity Hydrometers

Specific gravity hydrometer, 87 H - 62, 0,900 - 0,950 °SG, 0,005 °SG, 330 mm
  AMARH826510
 :   Amarell
Specific Gravity Hydrometers
  AMARH826510
 :  Amarell
Specific Gravity Hydrometers

Specific gravity hydrometer, 87 H - 62, 0,900 - 0,950 °SG, 0,005 °SG, 330 mm

 
59,10 €
   
1 * 1 ST
 

These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.

  • Capillary prismatic
  • Reliable and simple
  • Paper scale with type designation
  • Shot weighted body

 : Supplied in a transparent plastic case.

 
  • Description:
    Specific gravity hydrometer
  • Length:
    330 mm
  • Seq:
    32
  • Division:
    0,005 °SG
  • Model:
    87 H - 62
  • Range:
    0,900 - 0,950 °SG
  • Cat. No.:
    AMARH826510
  • Accuracy:
    ±1 scale division
  • Reference temperature:
    60/60 °F (except model AMARH826764-FL: 15 °C)