ContactUsLinkComponent
InfinityLab Quick Change valve head, 2 position /6 port, maximum pressure 800 bar, for use with multicolumn thermostat and 1290 Infi nity valve drive 1 * 1 items
Catalog # HEWL5067-4282
Supplier:  Agilent
InfinityLab Quick Change valve head, 2 position /6 port, maximum pressure 800 bar, for use with multicolumn thermostat and 1290 Infi nity valve drive 1 * 1 items
Catalog # HEWL5067-4282
Supplier:  Agilent
Frequently Bought Together