Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Cooling elements and frames

Cooling elements, −21 °C, Tempshell, Translucent white, −23...+19 °C, 275×155×60 mm
210×110×30 mm
Catalog # DLTTA100011
Supplier:   DELTA T
Cooling elements and frames
Catalog # DLTTA100011
Supplier:  DELTA T
Cooling elements and frames

Cooling elements, −21 °C, Tempshell, Translucent white, −23...+19 °C, 275×155×60 mm
210×110×30 mm

Price
141,00 €
Log in for availability
1 * 2 ST
Product Details & Documents

Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.

  • Made of PE and natural in colour
  • Sealing made of laminated aluminium film with a protected cap
  • A liquid crystal thermometer in the elements shows its current temperature

Used for transportation and storage of samples like blood products, homograft’s, cell cultures, stem cells, antibodies, proteins and other temperature sensitive medications.

Inside dimension: 210×110×30 mm

Specifications
  • Temperature Range:
    −23...+19 °C
  • Colour:
    Translucent white
  • Description:
    Cooling elements, −21 °C, Tempshell
  • Dimensions:
    275×155×60 mm
    210×110×30 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.