Order Entry
Austria
Orders LinkContactUsLinkComponent
Digital block heaters, Advanced | Avantor

Digital block heaters, Advanced

Digital HB4DG, with heat up time of 60 minutes (at 100 °C), 4, Ambient +5...120 °C, ±0,2 °C, 429×203×89 mm, 3,8 kg
Catalog # OHAU30392089
Supplier:   OHAUS
Digital block heaters, Advanced
Catalog # OHAU30392089
Supplier:  OHAUS
Digital block heaters, Advanced
Digital block heaters, Advanced

Digital HB4DG, with heat up time of 60 minutes (at 100 °C), 4, Ambient +5...120 °C, ±0,2 °C, 429×203×89 mm, 3,8 kg

Price
1 220,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Designed for applications that require repeatable results and superior temperature stability. These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures.

  • PID temperature control, with optional external RTD probe, offers a temperature stability as low as ±0,1 °C with a temperature uniformity as low as ±0,1 °C
  • Hot warning symbol light is illuminated when the temperature is above 40 °C
  • Overshoot protection ensures unit will automatically stop heating if the unit exceeds the set temperature by 10 °C
  • Audible alarm sounds when time reaches zero and when unit reaches set-point temperature
  • Temperature calibration mode allows user to calibrate unit to an external temperature device

Optional external temperature probe kit monitors actual block or sample temperature. Each model accepts separate interchangeable modular blocks, accommodating various tube sizes from 0,2 ml microtubes to 50 ml centrifuge tubes. Each block has a thermometer well for measuring block temperature. Efficient heating due to close tube and block contact for maximum heat retention.

Touch pad controls with easy to read, independent LED displays for temperature, and time. It provides repeatable and accurate results every time and is easily visible across lab benches. Shows set point and actual temperature. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.

Units can be run in conditions from 18 to 33 °C, 20 to 80% relative humidity, non-condensing.

Ordering information: Modular heating blocks must be ordered separately.

Caution: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.

Specifications
  • No. of blocks:
    4
  • Temperature Range:
    Ambient +5...120 °C
  • Weight:
    3,8 kg
  • Seq:
    03
  • W×D×H:
    429×203×89 mm
  • Temp. stability:
    ±0,2 °C
  • Type:
    Digital HB4DG, with heat up time of 60 minutes (at 100 °C)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Optional external temperature probe kit
    Optional external temperature probe kit
    OHAU30400246
    Log in for availability
    1 * 1 ST - 281,00 €

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.