pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 500 mL
Catalog # REGETB700
Supplier: REAGECON
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 500 mL
Catalog # REGETB700
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals