ContactUsLinkComponent
General Purpose Transfer Pipettes, Beral. Length 14,5 cm. Stem Diameter 2,8 mm. Overall Capacity 5,5 mL. Bulb Draw 2,4 mL. 22 drops per mL. 1 * 5.000 items
Catalog # SAMCB75-100
Supplier:  SAMCO
General Purpose Transfer Pipettes, Beral. Length 14,5 cm. Stem Diameter 2,8 mm. Overall Capacity 5,5 mL. Bulb Draw 2,4 mL. 22 drops per mL. 1 * 5.000 items
Catalog # SAMCB75-100
Supplier:  SAMCO
Frequently Bought Together