Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Mesitylene

Bottle, 500 mL
Catalog # APOSOR1559-500ML
Flammable
Irritant
Environment
Supplier:   Apollo Scientific
Mesitylene
Catalog # APOSOR1559-500ML
Supplier:  Apollo Scientific
CAS Number:  108-67-8
Mesitylene

Bottle, 500 mL

CAS Number:  108-67-8
Price
63,70 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Boiling Pt:163…166 °C
  • Melting Pt:–45 °C
  • Density:0.864 g/cm³ (25 °C)
  • Flash Pt:53 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.