Order Entry
Austria
Orders LinkContactUsLinkComponent
Mesitylene

Mesitylene

Bottle, 2,5 L
Catalog # APOSOR1559-2.5L
Flammable
Irritant
Environment
Supplier:   Apollo Scientific
Mesitylene
Catalog # APOSOR1559-2.5L
Supplier:  Apollo Scientific
CAS Number:  108-67-8
Mesitylene

Bottle, 2,5 L

CAS Number:  108-67-8
Price
258,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Boiling Pt:163…166 °C
  • Melting Pt:–45 °C
  • Density:0.864 g/cm³ (25 °C)
  • Flash Pt:53 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.