Order Entry
Austria
Orders LinkContactUsLinkComponent
Mesitylene

Mesitylene

Bottle, 100 mL
Catalog # APOSOR1559-100ML
Flammable
Irritant
Environment
Supplier:   Apollo Scientific
Mesitylene
Catalog # APOSOR1559-100ML
Supplier:  Apollo Scientific
CAS Number:  108-67-8
Mesitylene

Bottle, 100 mL

CAS Number:  108-67-8
Price
23,20 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Boiling Pt:163…166 °C
  • Melting Pt:–45 °C
  • Density:0.864 g/cm³ (25 °C)
  • Flash Pt:53 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.