Order Entry
Austria
Orders LinkContactUsLinkComponent
Copper(I) thiocyanate

Copper(I) thiocyanate

Bottle, 250 g
Catalog # APOSIN9847-250G
Irritant
Environment
Supplier:   Apollo Scientific
Copper(I) thiocyanate
Catalog # APOSIN9847-250G
Supplier:  Apollo Scientific
CAS Number:  1111-67-7
Copper(I) thiocyanate

Bottle, 250 g

CAS Number:  1111-67-7
Price
171,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide
  • CAS Number:1111-67-7
  • Formula:CCuNS
  • MDL Number:MFCD00010980
  • Molecular Weight:121.63 g/mol
  • Melting Pt:1084 °C
  • Density:2.84 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:214-183-1
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.