Order Entry
Austria
Orders LinkContactUsLinkComponent
Vancomycin hydrochloride

Vancomycin hydrochloride

Bottle, 1 g
Catalog # APOSBIV0155-1G
Reizend
Supplier:   Apollo Scientific
Vancomycin hydrochloride
Catalog # APOSBIV0155-1G
Supplier:  Apollo Scientific
CAS Number:  1404-93-9
Vancomycin hydrochloride

Bottle, 1 g

CAS Number:  1404-93-9
Price
29,00 €
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

Vancomycin is a glycopeptide antibiotic. It exerts its action by inhibiting the formation of the peptidoglycan polymers of the bacterial cell wall. Often used incombination with cefotaxim or carbenicillin.

  • Potency of 1099 mcg/mg
  • Cell culture tested.
Synonyme:  (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
  • CAS Nummer:1404-93-9
  • Formel:C₆₆H₇₅Cl₂N₉O₂₄·HCl
  • MDL-Nummer:MFCD03613611
  • Molekulargewicht:1485.71 g/mol
  • Lagertemperatur:Refrigerator
Spezifikationen

Spezifikationen

  • Pack type:
    Bottle
  • Pk:
    1 g

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.