Order Entry
Austria
Orders LinkContactUsLinkComponent
β-Cyclodextrine

β-Cyclodextrine

Bottle, 100 g
Catalog # APOSBIM1015-100G
Irritant
Supplier:   Apollo Scientific
β-Cyclodextrine
Catalog # APOSBIM1015-100G
Supplier:  Apollo Scientific
CAS Number:  7585-39-9
β-Cyclodextrine

Bottle, 100 g

CAS Number:  7585-39-9
Price
86,90 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Melting Pt:290…300 °C
  • Density:1.44 g/cm³ (25 °C)
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.