Order Entry
Austria
Orders LinkContactUsLinkComponent
 

D-(+)-Galactose

Bottle, 1 kg
Catalog # APOSBIG1724-1KG
Supplier:   Apollo Scientific
D-(+)-Galactose
Catalog # APOSBIG1724-1KG
Supplier:  Apollo Scientific
CAS Number:  59-23-4
D-(+)-Galactose

Bottle, 1 kg

CAS Number:  59-23-4
Price
86,90 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Melting Pt:168…170 °C
  • Density:1.5 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.