Order Entry
Austria
Orders LinkContactUsLinkComponent
Caffeic acid

Caffeic acid

Bottle, 100 g
Catalog # APOSBIB6021-100G
Irritant
Health
Supplier:   Apollo Scientific
Caffeic acid
Catalog # APOSBIB6021-100G
Supplier:  Apollo Scientific
CAS Number:  331-39-5
Caffeic acid

Bottle, 100 g

CAS Number:  331-39-5
Price
34,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Melting Pt:211…213 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-361-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.