L(+)-Ascorbic acid ≥99.5%
Bottle, 500 g
Artikelnr. APOSBIA0602-500G
Anbieter: Apollo Scientific
L(+)-Ascorbic acid ≥99.5%
Artikelnr. APOSBIA0602-500G
Anbieter: Apollo Scientific
CAS Nummer: 50-81-7
Springe zu:
Product Details & Documents
Synonyme: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Nummer:50-81-7
- Formel:C₆H₈O₆
- MDL-Nummer:MFCD00064328
- Molekulargewicht:176.13 g/mol
- Schmelzpunkt:190…194 °C (dec.)
- Dichte:1.65 g/cm³ (25 °C)
- Lagertemperatur:Ambient
- EINECS:200-066-2
Spezifikationen
Spezifikationen
- Pack type:Bottle
- Pk:500 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



