Order Entry
Austria
Orders LinkContactUsLinkComponent
Caffeic acid

Caffeic acid

Bottle, 25 g
Catalog # APOSBIB6021-25G
Irritant
Health
Supplier:   Apollo Scientific
Caffeic acid
Catalog # APOSBIB6021-25G
Supplier:  Apollo Scientific
CAS Number:  331-39-5
Caffeic acid

Bottle, 25 g

CAS Number:  331-39-5
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Melting Pt:211…213 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-361-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications