VWR®, Block Heaters, Digital
Product Details & Documents
Multi-purpose, high wattage, constant temperature, VWR® digital block heaters are economical, versatile and compact. These digital models provide reproducable results, exceptional temperature uniformity, and are ideal for a wide range of applications. Temperature calibration mode allows the user to calibrate the unit to an external temperature device. In timed mode, an alarm will sound when the time reaches zero and the unit reaches set point temperature.
- Compact design with small footprint
- Temperature range from ambient +5 to 120 °C
- 'Hot' symbol warning light is illuminated when the temperature is above 40 °C
- Accommodates VWR® interchangeable modular heating blocks for tubes from 0,2 to 50 ml capacity
- Can be used at 18 to 33 °C, with 20 to 80% relative humidity, non condensing
These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, denaturing proteins, DNA applications, ELISA, melting/boiling points, and a variety of other laboratory procedures.
Microprocessor control: PID microprocessor temperature control, with optional external RTD probe, offers a temperature stability as low as ±0,1 °C, with a temperature uniformity as low as ±0,1 °C. Samples are heated to temperature quickly and accurately, heat-up time to 100 ºC is 16 minutes. Temperature is adjusted in 0,1 °C increments.
LED display: Touch pad controls with easy to read, independent LED displays for temperature and time. Shows set point and actual temperatures. Timer displays elapsed time or, when programmed to user-defined limit, will shut off unit when time reaches zero.
Units have separate interchangeable modular heating blocks, which accommodate various sample containers such as microtubes, centrifuge tubes, vials, microplates and PCR strips or tubes. Blocks include a thermometer well, and feature close 'tube and block' contact for maximum heat retention. All dry block heaters include a built-in support rod holder for mounting the probe kit.
Certifications: TUV listed, RoHS compliant.
Ordering information: Supplied without heating blocks, please order these separately. Optional external temperature-probe kit monitors actual block or sample temperature.
Caution: To avoid a possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed for use as a dry bath/incubator only.
* RT = Ambient
Specifications
- Heating capacity:0,19 kW
- No. of blocks:2
- Temperature Range:RT* +5...120 °C
- Weight:2,6 kg
- Seq:002
- W×D×H:196×348×117 mm
- Temp. stability:±0,2 °C
- Type:Block heater, digital
- Cat. No.:460-0350
- Environmentally Preferable:Waste Reducing
Accessories
Product Information
- Catalog #Well sizeNo. of HolesAvailabilityPrice
- Single block, 2×6 cuvettes (12,5 mm)
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3243Ø 19,1 mm17/18 mm round bottom tubesLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block, 10×8-PCR tube stripsSingle block, 10×8-PCR tube strips460-3224Ø 6,4 mm0,2 ml tubesLog in for availability1 * 1 ST - 234,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeNo. of HolesAvailabilityPrice
- Single block, test tube combination 6 mm, 12/13 mm, 25 mmSingle block, test tube combination 6 mm, 12/13 mm, 25 mm460-3220Ø 8,3 / Ø 13,9 / Ø 26,2 mm6/5/3Log in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeNo. of HolesAvailabilityPrice
- Single block, microtube combination 0,5 ml, 1,5 ml, 2,0 mlSingle block, microtube combination 0,5 ml, 1,5 ml, 2,0 ml460-3248Ø 7,9 / Ø 11,1 / Ø 11,5 mm6/10/5Log in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3215Ø 13,9 mm12/13 mm round bottom tubesLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3212Ø 11,5 mm1,5 ml Eppendorf microtubesLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3216Ø 13,9 mm12/13 mm round bottom tubesLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeW×D×HAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeW×D×HAvailabilityPrice
- Double block
Product Information
- Catalog #Well sizeNo. of HolesAvailabilityPrice
- Single block, centrifuge tube combination 1,5 ml, 15 ml, 50 mlSingle block, centrifuge tube combination 1,5 ml, 15 ml, 50 ml460-3247Ø 11,1 / Ø 17,1 / Ø 29,0 mm4/3/2Log in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block, 64× individual PCR tubesSingle block, 64× individual PCR tubes460-3225Ø 6,4 mm0,2 ml tubesLog in for availability1 * 1 ST - 183,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3217Ø 17,5 mm15/16 mm round bottom tubesLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3234Ø 6,3 mmRound bottomed tubes, 6 mm ØLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Double block, 1×96-well PCR plateDouble block, 1×96-well PCR plate460-3226Ø 6,4 mm0,2 ml tubesLog in for availability1 * 1 ST - 409,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Double block
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single blockSingle block460-3245Ø 11,5 mm2,0 ml VWR/Eppendorf microtubesLog in for availability1 * 1 ST - 149,00 €
Product Information
- Catalog #Well sizeForAvailabilityPrice
- Single block
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



