ContactUsLinkComponent
SICCO-Maxi 1-Desiccator Black, PMMA/AL, B 560 mm x H 1150 mm x T 580 mm black acrylic glass/aluminium frame, with: elastic gasket-lined door, 1 desiccant tray, 1 mounted hygrometer, 1 magnetic lock, 4 removable shelves made of stainless steel, 4 cast 1 * 1 items
Catalog # BOHLV1936-07
Supplier:  Bohlender
undefined
SICCO-Maxi 1-Desiccator Black, PMMA/AL, B 560 mm x H 1150 mm x T 580 mm black acrylic glass/aluminium frame, with: elastic gasket-lined door, 1 desiccant tray, 1 mounted hygrometer, 1 magnetic lock, 4 removable shelves made of stainless steel, 4 cast 1 * 1 items
Catalog # BOHLV1936-07
Supplier:  Bohlender
Frequently Bought Together