Order Entry
Austria
Orders LinkContactUsLinkComponent
Hook connectors

Hook connectors

Hook connector, Stainless steel, 13 mm
Catalog # USBE2430NEU
Supplier:   USBECK Laborgeräte
Hook connectors
Catalog # USBE2430NEU
Hook connectors
Hook connectors

Hook connector, Stainless steel, 13 mm

Price
16,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Nickel plated brass screw with plastic handle.

  • For rods up to 13 mm
  • Angle: 90°
Specifications
  • Description:
    Hook connector
  • Material:
    Stainless steel
  • Seq:
    02
  • For rod Ø:
    13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.