ContactUsLinkComponent
1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride (EDC-HCl) 99 % Coupling reagent for peptide synthesis Empirical formula C8H17N3 . HCl Molar mass (M) 191,7 g/mol Melting point (mp)110-115 °C Solubility soluble (H2O, 20 °C) Storage temp.: - 1 * 5 g
Catalog # ROTH2156.1
Supplier:  Roth Carl
1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride (EDC-HCl) 99 % Coupling reagent for peptide synthesis Empirical formula C8H17N3 . HCl Molar mass (M) 191,7 g/mol Melting point (mp)110-115 °C Solubility soluble (H2O, 20 °C) Storage temp.: - 1 * 5 g
Catalog # ROTH2156.1
Supplier:  Roth Carl
Frequently Bought Together