Orders LinkContactUsLinkComponent
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
ESC swab Neutralising Rinse Solution, 100 tubes×10 ml
Catalog # 85601.
Supplier:   LIOFILCHEM
ESC (Easy Surface Checking) swabs
Catalog # 85601.
Supplier:  LIOFILCHEM
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
ESC swab Neutralising Rinse Solution, 100 tubes×10 ml
Price
146,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
  • Pre-filled swab device
  • Rayon bud
  • Wide range of diluents
  • Leak proof closure
  • Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).
Specifications
  • Description:
    ESC swab Neutralising Rinse Solution, 100 tubes×10 ml
  • Seq:
    13