Brushes, FlexBrush™
Microfibre cover (80% Polyester, 20% Polyamide)
Catalog # 141-1645
Supplier: MICRONOVA
Jump to:
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
- Bendable brush with polyester or microfibre cover
- Acme threaded end attaches to a variety of handles for extended reach
- Threaded handle (STA series) can be added for extended reach
- Covers are compatible with strong disinfectants and autoclavable
Specifications
- Description:Microfibre cover (80% Polyester, 20% Polyamide)
- Cat. No.:141-1645
- Environmentally Preferable:Waste Reducing



