300 ML REAGENT TROUGH SBS FORMAT BOTTOM WITH SQUARES 1 * 5 items
Catalog # HAMR56669-01
Supplier: HAMILTON ROBOTICS
300 ML REAGENT TROUGH SBS FORMAT BOTTOM WITH SQUARES 1 * 5 items
Catalog # HAMR56669-01
Supplier: HAMILTON ROBOTICS
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Pipette tips, Labtip
Supplier: Thermo Fisher Scientific
Water standards, CRM, Karl Fischer titration, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals